Search
Search

SLC5A11| Gene Symbol: SLC5A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501307.4 | XP_001501357.1 | 1349 | 79 | ||
| XM_014730049.2 | XP_014585535.1 | 1399 | 79 | |||
| XM_023616092.1 | XP_023471860.1 | 1293 | 79 | |||
| XM_023616093.1 | XP_023471861.1 | 1339 | 79 | |||
Set Membership: |
|