Search
Search

PRKCB| Gene Symbol: PRKCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616107.1 | XP_023471875.1 | 711 | 72 | ||
| XM_023616108.1 | XP_023471876.1 | 710 | 72 | |||
| XM_023616109.1 | XP_023471877.1 | 712 | 72 | |||
Set Membership: |
|