Search
Search
| Gene Symbol: NDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598888.3 | XP_005598945.2 | 913 | 65 | ||
| XM_023616274.1 | XP_023472042.1 | 648 | 65 | |||
| XM_023616275.1 | XP_023472043.1 | 717 | 65 | |||
| XM_023616276.1 | XP_023472044.1 | 648 | 65 | |||
| XM_023616277.1 | XP_023472045.1 | 830 | 65 | |||
Set Membership: |
|