Search
Search

CRAMP1| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730258.2 | XP_014585744.2 | 2557 | 95 | ||
| XM_023616636.1 | XP_023472404.1 | 1141 | 95 | |||
| XM_023616638.1 | XP_023472406.1 | 1360 | 95 | |||
Set Membership: |
|