Search
Search

RHOT2| Gene Symbol: RHOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616706.1 | XP_023472474.1 | 857 | 69 | ||
| XM_023616707.1 | XP_023472475.1 | 857 | 69 | |||
| XM_023616708.1 | XP_023472476.1 | 857 | 69 | |||
| XM_023616709.1 | XP_023472477.1 | 856 | 69 | |||
| XM_023616710.1 | XP_023472478.1 | 826 | 69 | |||
| XM_023616711.1 | XP_023472479.1 | 813 | 69 | |||
| XM_023616712.1 | XP_023472480.1 | 763 | 69 | |||
Set Membership: |
|