Search
Search
| Gene Symbol: DBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498360.5 | XP_001498410.4 | 988 | 74 | ||
| XM_023617106.1 | XP_023472874.1 | 387 | 74 | |||
| XM_023617107.1 | XP_023472875.1 | 383 | 74 | |||
| XM_023617108.1 | XP_023472876.1 | 283 | 74 | |||
| XM_023617109.1 | XP_023472877.1 | 465 | 74 | |||
Set Membership: |
|