Search
Search
| Gene Symbol: GRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730468.2 | XP_014585954.1 | 1087 | 63 | ||
| XM_023617110.1 | XP_023472878.1 | 1211 | 63 | |||
| XM_023617111.1 | XP_023472879.1 | 1259 | 63 | |||
| XM_023617112.1 | XP_023472880.1 | 1206 | 63 | |||
| XM_023617113.1 | XP_023472881.1 | 1175 | 63 | |||
| XM_023617114.1 | XP_023472882.1 | 1104 | 63 | |||
Set Membership: |
|