Search
Search

ZNF346| Gene Symbol: ZNF346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617150.1 | XP_023472918.1 | 614 | 94 | ||
| XM_023617151.1 | XP_023472919.1 | 612 | 94 | |||
| XM_023617152.1 | XP_023472920.1 | 565 | 94 | |||
Set Membership: |
|