Search
Search
| Gene Symbol: FAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502650.5 | XP_001502700.1 | 554 | 85 | ||
| XM_005599216.3 | XP_005599273.1 | 719 | 85 | |||
| XM_023617183.1 | XP_023472951.1 | 595 | 85 | |||