Search
Search

C14H5orf58| Gene Symbol: C14H5orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730531.2 | XP_014586017.1 | 238 | 104 | ||
| XM_014730532.2 | XP_014586018.1 | 234 | 104 | |||
| XM_023617262.1 | XP_023473030.1 | 131 | 104 | |||
| XM_023617263.1 | XP_023473031.1 | 237 | 104 | |||
Set Membership: |
|