Search
Search

PDGFRB| Gene Symbol: PDGFRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730580.2 | XP_014586066.2 | 1247 | 70 | ||
| XM_023617441.1 | XP_023473209.1 | 1292 | 70 | |||
| XM_023617442.1 | XP_023473210.1 | 280 | 70 | |||
Set Membership: |
|