Search
Search

ARHGEF37| Gene Symbol: ARHGEF37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501521.6 | XP_001501571.4 | 1756 | 94 | ||
| XM_023617446.1 | XP_023473214.1 | 1872 | 94 | |||
| XM_023617447.1 | XP_023473215.1 | 1876 | 94 | |||
| XM_023617448.1 | XP_023473216.1 | 2966 | 94 | |||
| XM_023617449.1 | XP_023473217.1 | 1879 | 94 | |||
| XM_023617450.1 | XP_023473218.1 | 2963 | 94 | |||
| XM_023617452.1 | XP_023473220.1 | 3614 | 94 | |||
| XM_023617453.1 | XP_023473221.1 | 3659 | 94 | |||
| XM_023617454.1 | XP_023473222.1 | 2843 | 94 | |||