Search
Search
| Gene Symbol: BRD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504292.5 | XP_001504342.1 | 191 | 64 | ||
| XM_005599382.3 | XP_005599439.1 | 337 | 64 | |||
| XM_005599383.3 | XP_005599440.1 | 191 | 64 | |||
| XM_014730648.2 | XP_014586134.1 | 592 | 64 | |||
| XM_014730649.2 | XP_014586135.1 | 337 | 64 | |||
| XM_014730650.2 | XP_014586136.1 | 337 | 64 | |||
| XM_014730651.2 | XP_014586137.1 | 337 | 64 | |||
| XM_014730652.2 | XP_014586138.1 | 143 | 64 | |||
| XM_023617631.1 | XP_023473399.1 | 337 | 64 | |||
Set Membership: |
|