Search
Search
| Gene Symbol: MYOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599387.2 | XP_005599444.1 | 631 | 67 | ||
| XM_005599388.3 | XP_005599445.1 | 406 | 67 | |||
| XM_014730661.2 | XP_014586147.1 | 982 | 67 | |||
| XM_023617646.1 | XP_023473414.1 | 383 | 67 | |||
Set Membership: |
|