Search
Search

CDKL3| Gene Symbol: CDKL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730677.2 | XP_014586163.1 | 1296 | 119 | ||
| XM_023617681.1 | XP_023473449.1 | 1254 | 119 | |||
| XM_023617682.1 | XP_023473450.1 | 1062 | 119 | |||
| XM_023617683.1 | XP_023473451.1 | 1056 | 119 | |||
| XM_023617684.1 | XP_023473452.1 | 541 | 119 | |||
| XM_023617685.1 | XP_023473453.1 | 1296 | 119 | |||
| XM_023617686.1 | XP_023473454.1 | 1296 | 119 | |||
Set Membership: |
|