Search
Search

SEPTIN8| Gene Symbol: SEPTIN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599435.3 | XP_005599492.2 | 293 | 77 | ||
| XM_005599436.3 | XP_005599493.2 | 291 | 77 | |||
| XM_014730685.2 | XP_014586171.2 | 294 | 77 | |||
| XM_023617709.1 | XP_023473477.1 | 295 | 77 | |||
| XM_023617710.1 | XP_023473478.1 | 240 | 77 | |||
| XM_023617711.1 | XP_023473479.1 | 295 | 77 | |||
| XM_023617712.1 | XP_023473480.1 | 297 | 77 | |||
Set Membership: |
|