Search
Search

KIF3A| Gene Symbol: KIF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502905.6 | XP_001502955.3 | 1816 | 88 | ||
| XM_003362820.4 | XP_003362868.1 | 1735 | 88 | |||
| XM_003362821.4 | XP_003362869.1 | 1744 | 88 | |||
| XM_023617714.1 | XP_023473482.1 | 1808 | 88 | |||
| GenBank mRNA | JL637101.1 | - | 2 - 3 | 251 | 88 | |
Set Membership: |
|