Search
Search

LYRM7| Gene Symbol: LYRM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362828.4 | XP_003362876.1 | 266 | 154 | ||
| XM_023617753.1 | XP_023473521.1 | 117 | 154 | |||
| GenBank mRNA | JL616256.2 | - | 2 - 3 | 127 | 154 | |
Set Membership: |
|