Search
Search

LOC100065733| Gene Symbol: LOC100065733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609888.3 | XP_005609945.1 | 1419 | 73 | ||
| XM_005609890.3 | XP_005609947.1 | 508 | 73 | |||
| XM_005609891.3 | XP_005609948.1 | 259 | 73 | |||
| GenBank mRNA | JN795138.1 | - | 1 - 2 | 87 | 73 | |
Set Membership: |
|