Search
Search

ZNF358| Gene Symbol: ZNF358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644624.1 | XP_023500392.1 | 331 | 100 | ||
| XM_023644625.1 | XP_023500393.1 | 782 | 100 | |||
| XM_023644626.1 | XP_023500394.1 | 748 | 100 | |||
| XM_023644627.1 | XP_023500395.1 | 482 | 100 | |||
| XM_023644628.1 | XP_023500396.1 | 241 | 100 | |||
Set Membership: |
|