Search
Search

FLYWCH2| Gene Symbol: FLYWCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498637.4 | XP_001498687.1 | 70 | 116 | ||
| XM_005599048.3 | XP_005599105.1 | 68 | 116 | |||
| XM_023616512.1 | XP_023472280.1 | 68 | 116 | |||
Set Membership: |
|