Search
Search

RELL2| Gene Symbol: RELL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599341.3 | XP_005599398.1 | 1366 | 101 | ||
| XM_023617525.1 | XP_023473293.1 | 1373 | 101 | |||
| XM_023617526.1 | XP_023473294.1 | 1122 | 101 | |||
| XM_023617527.1 | XP_023473295.1 | 1129 | 101 | |||
Set Membership: |
|