Search
Search

NOXRED1| Gene Symbol: NOXRED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605379.3 | XP_005605436.1 | 605 | 104 | ||
| XM_005605380.3 | XP_005605437.1 | 293 | 104 | |||
| XM_023628176.1 | XP_023483944.1 | 636 | 104 | |||
| XM_023628177.1 | XP_023483945.1 | 601 | 104 | |||
| XM_023628178.1 | XP_023483946.1 | 276 | 104 | |||
Set Membership: |
|