Search
Search

MAMSTR| Gene Symbol: MAMSTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729592.2 | XP_014585078.2 | 637 | 62 | ||
| XM_014729598.2 | XP_014585084.1 | 634 | 62 | |||
| XM_014729603.2 | XP_014585089.2 | 635 | 62 | |||
| XM_014729613.2 | XP_014585099.2 | 634 | 62 | |||
| XM_023650054.1 | XP_023505822.1 | 639 | 62 | |||
| XM_023650055.1 | XP_023505823.1 | 637 | 62 | |||
| XM_023650057.1 | XP_023505825.1 | 636 | 62 | |||
| XM_023650058.1 | XP_023505826.1 | 637 | 62 | |||
| XM_023650059.1 | XP_023505827.1 | 702 | 62 | |||
| XM_023650060.1 | XP_023505828.1 | 638 | 62 | |||
| XM_023650061.1 | XP_023505829.1 | 635 | 62 | |||
Set Membership: |
|