Search
Search

C2H8orf58| Gene Symbol: C2H8orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737753.2 | XP_014593239.1 | 2127 | 103 | ||
| XM_023636148.1 | XP_023491916.1 | 2105 | 103 | |||
| XM_023636149.1 | XP_023491917.1 | 1165 | 103 | |||
| XM_023636150.1 | XP_023491918.1 | 1917 | 103 | |||
| XM_023636151.1 | XP_023491919.1 | 1147 | 103 | |||
Set Membership: |
|