Search
Search

TULP2| Gene Symbol: TULP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489599.5 | XP_001489649.3 | 514 | 97 | ||
| XM_005596388.3 | XP_005596445.2 | 517 | 97 | |||
| XM_023650068.1 | XP_023505836.1 | 361 | 97 | |||
| XM_023650069.1 | XP_023505837.1 | 511 | 97 | |||
| XM_023650070.1 | XP_023505838.1 | 224 | 97 | |||
| XM_023650071.1 | XP_023505839.1 | 482 | 97 | |||
Set Membership: |
|