Search
Search

SYNE3| Gene Symbol: SYNE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628339.1 | XP_023484107.1 | 6066 | 78 | ||
| XM_023628340.1 | XP_023484108.1 | 954 | 78 | |||
| XM_023628341.1 | XP_023484109.1 | 1845 | 78 | |||
| XM_023628342.1 | XP_023484110.1 | 4262 | 78 | |||
| XM_023628343.1 | XP_023484111.1 | 6071 | 78 | |||