Search
Search

CACNA1F| Gene Symbol: CACNA1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633843.1 | XP_023489611.1 | 3833 | 79 | ||
| XM_023633845.1 | XP_023489613.1 | 3800 | 79 | |||
| XM_023633846.1 | XP_023489614.1 | 3638 | 79 | |||
Set Membership: |
|