Search
Search

FOXN4| Gene Symbol: FOXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646968.1 | XP_023502736.1 | 2058 | 68 | ||
| XM_023646969.1 | XP_023502737.1 | 2053 | 68 | |||
| XM_023646971.1 | XP_023502739.1 | 2052 | 68 | |||
| XM_023646972.1 | XP_023502740.1 | 2005 | 68 | |||
| XM_023646973.1 | XP_023502741.1 | 2001 | 68 | |||
| XM_023646974.1 | XP_023502742.1 | 1945 | 68 | |||
Set Membership: |
|