Search
Search

CHST9| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647597.1 | XP_023503365.1 | 339 | 93 | ||
| XM_023647598.1 | XP_023503366.1 | 427 | 93 | |||
| XM_023647599.1 | XP_023503367.1 | 2206 | 93 | |||
Set Membership: |
|