Search
Search

ETNPPL| Gene Symbol: ETNPPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503543.4 | XP_001503593.1 | 1068 | 60 | ||
| XM_005607898.3 | XP_005607955.1 | 1071 | 60 | |||
| XM_014737961.2 | XP_014593447.1 | 994 | 60 | |||