Search
Search
| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231900.5 | XP_001231901.4 | 19 - 20 | 5347 | 100 | |
| XM_015296621.2 | XP_015152107.1 | 19 - 20 | 5356 | 100 | ||
| XM_025143788.1 | XP_024999556.1 | 19 - 20 | 4717 | 100 | ||
| GenBank mRNA | AF465271.1 | - | 7 - 8 | 1364 | 100 | |
Set Membership: |
|