Search
Search

C1orf167| Gene Symbol: C1orf167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947511.3 | XP_004947568.1 | 3 - 4 | 298 | 122 | |
| XM_025142566.1 | XP_024998334.1 | 4 - 5 | 575 | 122 | ||
| XM_025142567.1 | XP_024998335.1 | 4 - 5 | 575 | 122 | ||
| GenBank mRNA | BX930872.1 | - | 2 - 3 | 226 | 122 | |
| BX932578.1 | - | 4 - 5 | 386 | 122 | ||
| BX934819.1 | - | 4 - 5 | 388 | 122 | ||
| BX950766.2 | - | 3 - 4 | 289 | 122 | ||
Set Membership: |
|