Search
Search

SLC16A6| Gene Symbol: SLC16A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279952.2 | XP_015135438.1 | 4 - 5 | 715 | 86 | |
| XM_015279953.2 | XP_015135439.1 | 2 - 3 | 384 | 86 | ||
| XM_415687.5 | XP_415687.2 | 4 - 5 | 736 | 86 | ||
| GenBank mRNA | BX933032.1 | - | 4 - 5 | 645 | 86 | |
| BX935693.2 | - | 4 - 5 | 644 | 86 | ||
Set Membership: |
|