Search
Search

PDCD5| Gene Symbol: PDCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277586.2 | NP_001264515.1 | 4 - 5 | 287 | 129 | |
| XM_015292431.2 | XP_015147917.1 | 4 - 5 | 443 | 129 | ||
| GenBank mRNA | BX933420.1 | - | 4 - 5 | 287 | 129 | |
| BX933425.1 | - | 4 - 5 | 280 | 129 | ||
| CR353650.1 | - | 2 - 3 | 777 | 129 | ||
Set Membership: |
|