Search
Search

NDUFS6| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419061.5 | XP_419061.1 | 3 - 4 | 343 | 73 | |
| GenBank mRNA | BX929731.1 | - | 670 | 73 | ||
| BX933021.1 | - | 3 - 4 | 333 | 73 | ||
| BX933813.1 | - | 3 - 4 | 333 | 73 | ||
| BX934156.1 | - | 3 - 4 | 311 | 73 | ||
| BX934371.1 | - | 3 - 4 | 322 | 73 | ||
Set Membership: |
|