Search
Search

EMX2| Gene Symbol: EMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288919.2 | XP_015144405.1 | 2 - 3 | 869 | 76 | |
| XM_025152057.1 | XP_025007825.1 | 2 - 3 | 605 | 76 | ||
| GenBank mRNA | BX934667.1 | - | 2 - 3 | 757 | 76 | |
Set Membership: |
|