Search
Search

BRD1| Gene Symbol: BRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199482.2 | NP_001186411.1 | 10 - 11 | 2854 | 66 | |
| XM_015302269.2 | XP_015157755.1 | 11 - 12 | 3621 | 66 | ||
| XM_025147458.1 | XP_025003226.1 | 11 - 12 | 3619 | 66 | ||
| XM_025147462.1 | XP_025003230.1 | 11 - 12 | 3236 | 66 | ||
| XM_025147464.1 | XP_025003232.1 | 11 - 12 | 3334 | 66 | ||
| XM_025147466.1 | XP_025003234.1 | 10 - 11 | 3132 | 66 | ||
| GenBank mRNA | CR352763.1 | - | 4 - 5 | 436 | 66 | |
Set Membership: |
|