Search
Search

CHODL| Gene Symbol: CHODL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231434.5 | XP_001231435.1 | 2 - 3 | 452 | 89 | |
| XM_004938355.3 | XP_004938412.1 | 2 - 3 | 511 | 89 | ||
| XM_004938357.3 | XP_004938414.1 | 2 - 3 | 511 | 89 | ||
| XM_004938358.3 | XP_004938415.1 | 2 - 3 | 512 | 89 | ||
| XM_004938359.3 | XP_004938416.1 | 2 - 3 | 511 | 89 | ||
| XM_004938360.3 | XP_004938417.1 | 2 - 3 | 511 | 89 | ||
| XM_416682.6 | XP_416682.2 | 2 - 3 | 494 | 89 | ||
| GenBank mRNA | BX931378.1 | - | 2 - 3 | 477 | 89 | |
| BX933424.2 | - | 2 - 3 | 477 | 89 | ||
| BX934063.2 | - | 1 - 2 | 194 | 89 | ||
Set Membership: |
|