Search
Search

COQ7| Gene Symbol: COQ7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277172.1 | NP_001264101.1 | 2 - 3 | 294 | 140 | |
| XM_015294222.2 | XP_015149708.1 | 2 - 3 | 358 | 140 | ||
| XM_015294223.2 | XP_015149709.1 | 2 - 3 | 358 | 140 | ||
| XM_025155105.1 | XP_025010873.1 | 2 - 3 | 358 | 140 | ||
| XM_025155106.1 | XP_025010874.1 | 2 - 3 | 358 | 140 | ||
| GenBank mRNA | BX933254.1 | - | 2 - 3 | 292 | 140 | |
| BX935091.1 | - | 2 - 3 | 289 | 140 | ||
| BX935416.1 | - | 2 - 3 | 299 | 140 | ||
| CR385437.1 | - | 2 - 3 | 294 | 140 | ||
Set Membership: |
|