Search
Search

PPDPF| Gene Symbol: PPDPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197037.1 | NP_001183966.1 | 3 - 4 | 306 | 121 | |
| GenBank mRNA | BX930759.1 | - | 3 - 4 | 297 | 121 | |
| BX931456.2 | - | 3 - 4 | 297 | 121 | ||
| BX935797.2 | - | 3 - 4 | 293 | 121 | ||
| BX935801.1 | - | 3 - 4 | 301 | 121 | ||
| BX935878.2 | - | 3 - 4 | 276 | 121 | ||
| BX936190.2 | - | 3 - 4 | 302 | 121 | ||
Set Membership: |
|