Search
Search

TDP1| Gene Symbol: TDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151003.1 | XP_025006771.1 | 5 - 6 | 894 | 68 | |
| XM_421313.6 | XP_421313.1 | 5 - 6 | 814 | 68 | ||
| GenBank mRNA | CR385144.1 | - | 4 - 5 | 246 | 68 | |
| CR387750.1 | - | 4 - 5 | 760 | 68 | ||
Set Membership: |
|