Search
Search

RTTN| Gene Symbol: RTTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256543.1 | NP_001243472.1 | 43 - 44 | 5835 | 92 | |
| XM_015282392.2 | XP_015137878.1 | 43 - 44 | 5866 | 92 | ||
| XM_025147287.1 | XP_025003055.1 | 42 - 43 | 5751 | 92 | ||
| XM_025147289.1 | XP_025003057.1 | 34 - 35 | 4648 | 92 | ||
| XM_025147290.1 | XP_025003058.1 | 27 - 28 | 3535 | 92 | ||
| GenBank mRNA | BX929955.2 | - | 1 - 2 | 67 | 92 | |
| CR389291.1 | - | 10 - 11 | 1273 | 92 | ||
Set Membership: |
|