Search
Search

DGKI| Gene Symbol: DGKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286157.2 | XP_015141643.2 | 24 - 25 | 2210 | 88 | |
| XM_025156046.1 | XP_025011814.1 | 24 - 25 | 2195 | 88 | ||
| XM_025156048.1 | XP_025011816.1 | 23 - 24 | 2144 | 88 | ||
| XM_025156051.1 | XP_025011819.1 | 23 - 24 | 2113 | 88 | ||
| XM_025156054.1 | XP_025011822.1 | 23 - 24 | 2098 | 88 | ||
| XM_025156057.1 | XP_025011825.1 | 22 - 23 | 2047 | 88 | ||
| XM_025156061.1 | XP_025011829.1 | 24 - 25 | 2498 | 88 | ||
| GenBank mRNA | CR389760.1 | - | 3 - 4 | 324 | 88 | |
Set Membership: |
|