Search
Search

CEP85| Gene Symbol: CEP85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947671.3 | XP_004947728.1 | 6 - 7 | 1488 | 85 | |
| XM_015297564.2 | XP_015153050.1 | 6 - 7 | 1420 | 85 | ||
| XM_015297565.2 | XP_015153051.1 | 6 - 7 | 2017 | 85 | ||
| XM_417697.6 | XP_417697.2 | 8 - 9 | 1571 | 85 | ||
| GenBank mRNA | CR389810.1 | - | 2 - 3 | 363 | 85 | |
Set Membership: |
|