Search
Search

ERI3| Gene Symbol: ERI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291303.1 | XP_015146789.1 | 4 - 5 | 480 | 55 | |
| XM_015291304.2 | XP_015146790.1 | 4 - 5 | 465 | 55 | ||
| XM_015291305.2 | XP_015146791.1 | 4 - 5 | 584 | 55 | ||
| GenBank mRNA | CR389892.1 | - | 3 - 4 | 455 | 55 | |
Set Membership: |
|