Search
Search

TPRN| Gene Symbol: TPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415534.6 | XP_415534.6 | 3 - 4 | 1272 | 73 | |
| GenBank mRNA | CR389919.1 | - | 2 - 3 | 180 | 73 | |
| CR406013.1 | - | 2 - 3 | 180 | 73 | ||
Set Membership: |
|