Search
Search

FIGNL1| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234038.5 | XP_001234039.1 | 2 | 2244 | 154 | |
| XM_004935143.3 | XP_004935200.1 | 2 | 2390 | 154 | ||
| GenBank mRNA | CR391400.1 | - | 1 | 1450 | 151 | |
Set Membership: |
|