Search
Search

STKLD1| Gene Symbol: STKLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945935.3 | XP_004945992.1 | 16 - 17 | 2266 | 76 | |
| XM_004945936.3 | XP_004945993.1 | 16 - 17 | 2248 | 76 | ||
| XM_004945937.3 | XP_004945994.1 | 15 - 16 | 2162 | 76 | ||
| XM_004945938.3 | XP_004945995.1 | 15 - 16 | 2135 | 76 | ||
| XM_004945939.3 | XP_004945996.1 | 16 - 17 | 1986 | 76 | ||
| XM_025141407.1 | XP_024997175.1 | 15 - 16 | 2116 | 76 | ||
| XM_025141408.1 | XP_024997176.1 | 16 - 17 | 1980 | 76 | ||
| GenBank mRNA | CR523822.1 | - | 5 - 6 | 621 | 76 | |
Set Membership: |
|