Search
Search

BBS5| Gene Symbol: BBS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252031.2 | NP_001238960.1 | 10 - 11 | 923 | 71 | |
| XM_025152192.1 | XP_025007960.1 | 10 - 11 | 1172 | 71 | ||
| XM_025152193.1 | XP_025007961.1 | 10 - 11 | 917 | 71 | ||
| GenBank mRNA | CR407505.1 | - | 10 - 11 | 923 | 71 | |
Set Membership: |
|